missing translation for 'onlineSavingsMsg'
Learn More

Adenine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019210
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019210 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019210 Supplier Merck Emd Millipore Supplier No. PHR13831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Adenine is a purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro. It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 73-24-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C5H5N5
MDL Number MFCD00041790
Melting Point >360°C
Synonym Adenine; 6-Aminopurine; Vitamin B4
Type Certified Reference Material
Quantity 1 g
Formula Weight 135.13
Grade Certified Reference Material
Show More Show Less