missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hydrochloride Hemihydrate 98.0+%, TCI America™

Supplier:  TCI America A01505G

 View more versions of this product

Catalog No. A01505G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Adenine Hydrochloride Hemihydrate
White
MFCD00038990
adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
Cl.NC1=C2NC=NC2=NC=N1
171.59
171.59
Crystalline Powder
2922-28-3
C5H6ClN5
5 g
UQVDQSWZQXDUJB-UHFFFAOYSA-N
7H-purin-6-amine hydrochloride
76219
≥98.0% (T)

Chemical Identifiers

2922-28-3
171.59
UQVDQSWZQXDUJB-UHFFFAOYSA-N
76219
Cl.NC1=C2NC=NC2=NC=N1
C5H6ClN5
MFCD00038990
adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
7H-purin-6-amine hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0045

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions