missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hydrochloride Hemihydrate 98.0+%, TCI America™

Catalog No. p-7116741
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01505G 5 g
A015025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01505G Supplier TCI America Supplier No. A01505G

Chemical Identifiers

CAS 2922-28-3
Molecular Formula C5H6ClN5
Molecular Weight (g/mol) 171.59
MDL Number MFCD00038990
InChI Key UQVDQSWZQXDUJB-UHFFFAOYSA-N
Synonym adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
PubChem CID 76219
IUPAC Name 7H-purin-6-amine hydrochloride
SMILES Cl.NC1=C2NC=NC2=NC=N1

Specifications

Color White
Quantity 5 g
Formula Weight 171.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Adenine Hydrochloride Hemihydrate
EINECSNumber (9)-0045
TSCA Yes