missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hemisulfate Dihydrate, ≥99%, MP Biomedicals™

Catalog No. p-4679546
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10019501 1 g
ICN10019580 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10019501 Supplier MP Biomedicals Inc Supplier No. 0210019501
Only null left

A purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro; It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP)

Chemical Identifiers

CAS 6509-19-9
Molecular Formula C10H16N10O6S
Molecular Weight (g/mol) 404.36
MDL Number MFCD00150149
InChI Key MKPCNMXYTMQZBE-UHFFFAOYSA-N
Synonym 9h-purin-6-amine hemisulfate dihydrate, adenine hemisulfate dihydrate, 6-aminopurine sulfate dihydrate, bis ade sulfuric acid dihydrate, sulfuric acid bis vitamin b4 dihydrate, bis adenine sulfuric acid dihydrate, sulfuric acid-7h-purin-6-amine-water 1/2/2
PubChem CID 44134512
IUPAC Name bis(7H-purin-6-amine) sulfuric acid dihydrate
SMILES O.O.OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1

Specifications

Color White
Assay Percent Range ≥99%
Quantity 1 g
Formula Weight 404.37
Physical Form Solid
Chemical Name or Material Adenine Hemisulfate Dihydrate
EINECSNumber 206-286-5
Recommended Storage Store at room temperature.