missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adefovir Dipivoxil 98.0+%, TCI America™
$63.52 - $369.64
Chemical Identifiers
| CAS | 142340-99-6 |
|---|---|
| Molecular Formula | C20H32N5O8P |
| Molecular Weight (g/mol) | 501.48 |
| MDL Number | MFCD00869897 |
| InChI Key | WOZSCQDILHKSGG-UHFFFAOYSA-N |
| Synonym | 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine |
| PubChem CID | 60871 |
| IUPAC Name | [({[2-(6-amino-9H-purin-9-yl)ethoxy]methyl}({[(2,2-dimethylpropanoyl)oxy]methoxy})phosphoryl)oxy]methyl 2,2-dimethylpropanoate |
| SMILES | CC(C)(C)C(=O)OCOP(=O)(COCCN1C=NC2=C(N)N=CN=C12)OCOC(=O)C(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2414100MG
|
TCI America
A2414100MG |
100 mg |
Each for $63.52
|
|
|||||
|
A24141G
|
TCI America
A24141G |
1 g |
Each for $369.64
|
|
|||||
Chemical Identifiers
| 142340-99-6 | |
| 501.48 | |
| WOZSCQDILHKSGG-UHFFFAOYSA-N | |
| 60871 | |
| CC(C)(C)C(=O)OCOP(=O)(COCCN1C=NC2=C(N)N=CN=C12)OCOC(=O)C(C)(C)C |
| C20H32N5O8P | |
| MFCD00869897 | |
| 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine | |
| [({[2-(6-amino-9H-purin-9-yl)ethoxy]methyl}({[(2,2-dimethylpropanoyl)oxy]methoxy})phosphoryl)oxy]methyl 2,2-dimethylpropanoate |
Specifications
| 142340-99-6 | |
| White-Yellow | |
| MFCD00869897 | |
| 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine | |
| CC(C)(C)C(=O)OCOP(=O)(COCCN1C=NC2=C(N)N=CN=C12)OCOC(=O)C(C)(C)C | |
| 501.48 | |
| 501.48 | |
| Crystalline Powder |
| 101°C | |
| C20H32N5O8P | |
| 100 mg | |
| WOZSCQDILHKSGG-UHFFFAOYSA-N | |
| [({[2-(6-amino-9H-purin-9-yl)ethoxy]methyl}({[(2,2-dimethylpropanoyl)oxy]methoxy})phosphoryl)oxy]methyl 2,2-dimethylpropanoate | |
| 60871 | |
| ≥98.0% (HPLC,T) | |
| Adefovir Dipivoxil |
Safety and Handling
RTECSNumber : UA2459362
TSCA : No
Recommended Storage : Freezer