accessibility menu, dialog, popup

UserName

CAS RN 142340-99-6

H3CCH3H3COOOPOONNNH2NNOOOCH3H3CH3C

CAS RN 142340-99-6

IUPAC Name: [({[2-(6-amino-9H-purin-9-yl)ethoxy]methyl}({[(2,2-dimethylpropanoyl)oxy]methoxy})phosphoryl)oxy]methyl 2,2-dimethylpropanoate
Synonyms: adefovir dipivoxil preveon adefovir dipivoxil preparation adefovir depivoxil ADV adefovir dipivoxil (substance) PMEA hepsera adefovir pivoxil gilead brand of adefovir dipivoxil
Molecular Weight (g/mol): 501.48
Molecular Formula: C20H32N5O8P
InChi Key: WOZSCQDILHKSGG-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)OCOP(=O)(COCCN1C=NC2=C(N)N=CN=C12)OCOC(=O)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results