Learn More
Promega ADCC Reporter Bioassay, V Variant Core Kit
Catalog No. PRG7018
Encompass
Change view
Click to view available options
Quantity:
1 of each component
5 of each component
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG7018 5 of each component
PRG7010 1 of each component
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG7018 Supplier Promega Supplier No. G7018
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
  • RPMI 1640 Medium
  • Low IgG Serum
  • Bio-Glo™ Luciferase Assay Buffer
  • Bio-Glo™ Luciferase Assay Substrate
  • ADCC Bioassay Effector Cells
Provides a biologically relevant and specific mechanism-of-action-based measure of ADCC without the complex workflow and variability inherent in primary cell-based assays. The bioassay also can be used to quantify effects of antibody glycosylation on Fc effector function.
Specifications
Content And Storage Varies Among Components
Quantity 5 of each component