Learn More
Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 4-Place Rotor with Rectangular Bucket
Adapters are sold individually and are available in other sizes for convience.
$262.47 - $394.68
| Catalog Number | Mfr. No. | Container Type | Color | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Container Type | Color | Price | Quantity & Availability | ||||
|
13100608
|
Fisherbrand™
75005327 |
Bottle | White |
Each for $272.96
|
|
||||
|
13100607
|
Fisherbrand™
75005387 |
Bottle | Brown |
Each for $394.68
|
|
||||
|
13100609
|
Fisherbrand™
75005330 |
Bottle | Gray |
Each for $262.47
|
|
||||
Description
Fit in rectangular bucket 13-100-587 for swinging bucket rotor 13-100-584; also can be used in sealed rotor 13-100-589.
Order Info
Adapters are sold individually. Order correct quantity to outfit rotor. Other adapter sizes are available online.
Specifications
| accuSpin 1/1R 4-Place Rotor |