missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 4-Place Rotor with Rectangular Bucket

Catalog No. 13100608
Change view
Click to view available options
Color:
Brown
Gray
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13100608 White
13100607 Brown
13100609 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13100608 Supplier Fisherbrand™ Supplier No. 75005327

Adapters are sold individually and are available in other sizes for convience.

Fit in rectangular bucket 13-100-587 for swinging bucket rotor 13-100-584; also can be used in sealed rotor 13-100-589.

Order Info

Adapters are sold individually. Order correct quantity to outfit rotor. Other adapter sizes are available online.

Specifications

For Use With (Equipment) accuSpin 1/1R 4-Place Rotor
Color White