missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapter
Adapter for Rotor A-4-81 and Rotor S-4x500.
Supplier: Eppendorf™ 022638707
Description
- For round-bottom tubes
- For 500 mL rectangular bucket in Rotor A-4-81 and Rotor S-4x500
- 2 pcs.
Specifications
| Adapter | |
| Rotor A-4-81 and Rotor S-4x500 | |
| 1.2 to 5 mL |
| Round Bottom | |
| 20 |