missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Adapalene

Catalog No. AC464500010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464500010 1 g
AC464500050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464500010 Supplier Thermo Scientific™ Supplier No. 464500010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 106685-40-9
Molecular Formula C28H28O3
Molecular Weight (g/mol) 412.53
InChI Key LZCDAPDGXCYOEH-UHFFFAOYSA-N
IUPAC Name 6-[3-(adamantan-1-yl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
SMILES COC1=CC=C(C=C1C12CC3CC(CC(C3)C1)C2)C1=CC=C2C=C(C=CC2=C1)C(O)=O

Specifications

Quantity 1 g
Formula Weight 412.53
Chemical Name or Material Adapalene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only