missing translation for 'onlineSavingsMsg'
Learn More

Adamantane 99.0+%, TCI America™

Supplier:  TCI America A069625G

 View more versions of this product

Catalog No. A069625G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Adamantane
White
MFCD00074719
tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl
C1C2CC3CC1CC(C2)C3
136.238
CHEBI:40519
≥99.0% (GC)
281-23-2
C10H16
25 g
ORILYTVJVMAKLC-UHFFFAOYSA-N
adamantane
9238
136.24
Crystalline Powder

Chemical Identifiers

281-23-2
136.238
ORILYTVJVMAKLC-UHFFFAOYSA-N
9238
adamantane
C10H16
MFCD00074719
tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl
CHEBI:40519
C1C2CC3CC1CC(C2)C3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0655

RTECSNumber : YK0493000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions