missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1-carbonyl chloride, 97%

Catalog No. AAL0235218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0235218 50 g
AAL0235209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0235218 Supplier Thermo Scientific Chemicals Supplier No. L0235218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2094-72-6 | C11H15ClO | 198.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2094-72-6
Molecular Formula C11H15ClO
Molecular Weight (g/mol) 198.69
MDL Number MFCD00074724
InChI Key MIBQYWIOHFTKHD-UHFFFAOYSA-N
Synonym 1-adamantanecarbonyl chloride, adamantanecarbonyl chloride, 1-adamantanecarbonylchloride, 1-adamantoyl chloride, adamantoyl chloride, 1-adamantanoic acid chloride, 1-adamantanecarboxylic acid chloride, adamantane-1-carboxylic acid chloride, 1-adamantylcarbonyl chloride, 1-adamantyl carbonyl chloride
PubChem CID 98915
IUPAC Name adamantane-1-carbonyl chloride
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)Cl

Specifications

Melting Point 48°C to 53°C
Boiling Point 135°C to 136°C (10 mmHg)
Quantity 50 g
UN Number UN3261
Beilstein 389960
Sensitivity Moisture sensitive
Formula Weight 198.69
Percent Purity 97%
Chemical Name or Material Adamantane-1-carbonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-252-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only