accessibility menu, dialog, popup

UserName

CAS RN 2094-72-6

IUPAC Name: adamantane-1-carbonyl chloride
Synonyms: adamantane-1-carbonyl chloride
Molecular Weight (g/mol): 198.69
Molecular Formula: C11H15ClO
InChi Key: MIBQYWIOHFTKHD-UHFFFAOYSA-N
SMILES: ClC(=O)C12CC3CC(CC(C3)C1)C2
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)