missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAM8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5149786

Catalog No. PA5149786


Only null left

Description

Description

ADAM8 Polyclonal Antibody for Western Blot, ICC/IF, IHC

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene may be involved in cell adhesion during neurodegeneration, and it is thought to be a target for allergic respiratory diseases, including asthma. Alternative splicing results in multiple transcript variants.
Specifications

Chemical Identifiers

ADAM8
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P78325, Q05910
101, 11501, 499285
Recombinant A Disintegrin And Metalloprotease 8 (ADAM8) (Glu145-Cys493).
Antibody
0.5 mg/mL
Immunohistochemistry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
ADAM8
a disintegrin and metallopeptidase domain 8; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 8; ADAM; ADAM 8; ADAM metallopeptidase domain 8; Adam8; ADAMs; CD156; CD156a; Cell surface antigen MS2; disintegrin and metalloproteinase domain-containing protein 8; E430039A18Rik; human leukocyte differentiation antigen; Macrophage cysteine-rich glycoprotein; metalloendopeptidases; MGC134985; MS2; RGD1566394
ADAM8
Primary
Protein A

Specifications

ADAM8
Polyclonal
Unconjugated
ADAM8
a disintegrin and metallopeptidase domain 8; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 8; ADAM; ADAM 8; ADAM metallopeptidase domain 8; Adam8; ADAMs; CD156; CD156a; Cell surface antigen MS2; disintegrin and metalloproteinase domain-containing protein 8; E430039A18Rik; human leukocyte differentiation antigen; Macrophage cysteine-rich glycoprotein; metalloendopeptidases; MGC134985; MS2; RGD1566394
Rabbit
Protein A
RUO
101, 11501, 499285
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P78325, Q05910
ADAM8
Recombinant A Disintegrin And Metalloprotease 8 (ADAM8) (Glu145-Cys493).
200 μL
Primary
Human, Mouse, Rat
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions