missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150213
Description
ADAM5 Polyclonal Antibody for Western Blot, ICC/IF, IHC
ADAM5 (putative disintegrin and metalloproteinase domain-containing protein 5) is also named as ADAM5P, TMDC2(tMDCII). The tMDC II, abundantly expressed, sperm surface MDC proteins identified in rodent and macaque systems, is also nonfunctional in the human. It has 7 isoforms produced by alternative splicing. It is expressed in testis highly by northern blot.Chemical Identifiers
| ADAM5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5BK84 | |
| 498654 | |
| Recombinant A Disintegrin And Metalloprotease 5 (ADAM5) (Cys413-Gln649). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| ADAM5 | |
| ADAM metallopeptidase domain 5 (pseudogene); ADAM5; ADAM5P; TMDCII | |
| ADAM5 | |
| Primary | |
| Protein A |
Specifications
| ADAM5 | |
| Polyclonal | |
| Unconjugated | |
| ADAM5 | |
| ADAM metallopeptidase domain 5 (pseudogene); ADAM5; ADAM5P; TMDCII | |
| Rabbit | |
| Protein A | |
| RUO | |
| 498654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5BK84 | |
| ADAM5 | |
| Recombinant A Disintegrin And Metalloprotease 5 (ADAM5) (Cys413-Gln649). | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody |