Learn More
Fisherbrand™ ADA Height Wood Knee Space Panels for use with Apron Rails
Fisherbrand™ Wood Knee Space Panels bring quality, warmth and durability to your laboratory.
 View more versions of this product
Catalog No. FB902448O1
Only null left
Explore available promotions
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Knee space panels close off the casework to the wall to create a clear sitting or standing space under the work surface.
Compliance
SEFA 8
Specifications
Specifications
Natural Stain
Apron Rails
2.5 cm
82.6 cm
30.5 to 121.9 cm
Wood (Oak Veneer)
1 in.
32.5 in.
12 to 48 in.
Product Suggestions
Product Suggestions