Learn More
Thermo Scientific Chemicals ADA, 98+%
CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol
$68.33 - $813.81
Chemical Identifiers
| CAS | 26239-55-4 |
|---|---|
| Molecular Formula | C6H10N2O5 |
| Molecular Weight (g/mol) | 190.155 |
| MDL Number | MFCD00008031 |
| InChI Key | QZTKDVCDBIDYMD-UHFFFAOYSA-N |
| Synonym | n-2-acetamido iminodiacetic acid, 2,2'-2-amino-2-oxoethyl azanediyl diacetic acid, carbamoylmethylaminodiacetic acid, ada, 2,2'-2-amino-2-oxoethyl imino diacetic acid, n-carbamoylmethyl iminodiacetic acid, glycine, n-2-amino-2-oxoethyl-n-carboxymethyl, n-carbamoylmethyliminodi acetic acid, unii-5c4r3o704e |
| PubChem CID | 117765 |
| ChEBI | CHEBI:43960 |
| IUPAC Name | 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid |
| SMILES | C(C(=O)N)N(CC(=O)O)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1526714
|
Thermo Scientific Chemicals
A1526714 |
25 g |
N/A
|
|
|||||
|
AAA1526722
|
Thermo Scientific Chemicals
A1526722 |
100 g |
N/A
|
|
|||||
|
AAA1526736
|
Thermo Scientific Chemicals
A1526736 |
500 g |
N/A
|
|
|||||
Description
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 26239-55-4 | |
| 190.155 | |
| QZTKDVCDBIDYMD-UHFFFAOYSA-N | |
| 117765 | |
| 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid |
| C6H10N2O5 | |
| MFCD00008031 | |
| n-2-acetamido iminodiacetic acid, 2,2'-2-amino-2-oxoethyl azanediyl diacetic acid, carbamoylmethylaminodiacetic acid, ada, 2,2'-2-amino-2-oxoethyl imino diacetic acid, n-carbamoylmethyl iminodiacetic acid, glycine, n-2-amino-2-oxoethyl-n-carboxymethyl, n-carbamoylmethyliminodi acetic acid, unii-5c4r3o704e | |
| CHEBI:43960 | |
| C(C(=O)N)N(CC(=O)O)CC(=O)O |
Specifications
| 26239-55-4 | |
| C6H10N2O5 | |
| 25 g | |
| 14,147 | |
| Insoluble in water. | |
| C(C(=O)N)N(CC(=O)O)CC(=O)O | |
| 190.155 | |
| CHEBI:43960 | |
| ≥98% |
| ∼218°C (decomposition) | |
| MFCD00008031 | |
| 1787181 | |
| n-2-acetamido iminodiacetic acid, 2,2'-2-amino-2-oxoethyl azanediyl diacetic acid, carbamoylmethylaminodiacetic acid, ada, 2,2'-2-amino-2-oxoethyl imino diacetic acid, n-carbamoylmethyl iminodiacetic acid, glycine, n-2-amino-2-oxoethyl-n-carboxymethyl, n-carbamoylmethyliminodi acetic acid, unii-5c4r3o704e | |
| QZTKDVCDBIDYMD-UHFFFAOYSA-N | |
| 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid | |
| 117765 | |
| 190.16 | |
| ADA |
Safety and Handling
EINECSNumber : 247-530-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only