missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ADA, 98+%

Catalog No. aaa1526722
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1526722 100 g
AAA1526714 25 g
AAA1526736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1526722 Supplier Thermo Scientific Chemicals Supplier No. A1526722
Only null left

CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol

ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 26239-55-4
Molecular Formula C6H10N2O5
Molecular Weight (g/mol) 190.155
MDL Number MFCD00008031
InChI Key QZTKDVCDBIDYMD-UHFFFAOYSA-N
Synonym n-2-acetamido iminodiacetic acid, 2,2'-2-amino-2-oxoethyl azanediyl diacetic acid, carbamoylmethylaminodiacetic acid, ada, 2,2'-2-amino-2-oxoethyl imino diacetic acid, n-carbamoylmethyl iminodiacetic acid, glycine, n-2-amino-2-oxoethyl-n-carboxymethyl, n-carbamoylmethyliminodi acetic acid, unii-5c4r3o704e
PubChem CID 117765
ChEBI CHEBI:43960
IUPAC Name 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)O

Specifications

Melting Point ∼218°C (decomposition)
Quantity 100 g
Beilstein 1787181
Merck Index 14,147
Solubility Information Insoluble in water.
Formula Weight 190.16
Percent Purity ≥98%
Chemical Name or Material ADA
EINECSNumber 247-530-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only