missing translation for 'onlineSavingsMsg'
Learn More

Medisca Acyclovir USP

Catalog No. 14185618
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185618 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185618 Supplier Medisca Supplier No. 017308
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acyclovir USP is a white to off-white crystalline powder. Alternative name(s): Aciclovir

Acyclovir USP is a white to off-white crystalline powder.

Alternative name(s): Aciclovir

Specifications

Chemical Name or Material Acyclovir USP
Packaging Jar 1500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT
CAS 59277-89-3
Type Drug Standard
Molecular Weight (g/mol) 225.2
Quantity 500 g
Percent Purity 98 to 102 %
Grade USP