missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Acyclovir | |
| 262°C | |
| C8H11N5O3 | |
| 1 g | |
| MKUXAQIIEYXACX-UHFFFAOYSA-N | |
| 2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-1H-purin-6-one | |
| 2022 | |
| 225.21 | |
| Crystalline Powder |
| 59277-89-3 | |
| White | |
| MFCD00057880 | |
| acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum | |
| NC1=NC2=C(N=CN2COCCO)C(=O)N1 | |
| 225.21 | |
| CHEBI:2453 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 59277-89-3 | |
| 225.21 | |
| MKUXAQIIEYXACX-UHFFFAOYSA-N | |
| 2022 | |
| 2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-1H-purin-6-one |
| C8H11N5O3 | |
| MFCD00057880 | |
| acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum | |
| CHEBI:2453 | |
| NC1=NC2=C(N=CN2COCCO)C(=O)N1 |
Safety and Handling
RTECSNumber : UP0791400
TSCA : No