Learn More
Acycloguanosine, 98%
Inhibits cytomegalovirus replication; induces apoptosis | CAS: 59277-89-3 | C8H11N5O3 | 225.21 g/mol
Supplier: Thermo Scientific Chemicals J6414406
Description
Inhibits cytomegalovirus replication; induces apoptosis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acycloguanosine | |
| 256°C to 260°C | |
| C8H11N5O3 | |
| 5 g | |
| acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum | |
| MKUXAQIIEYXACX-UHFFFAOYSA-N | |
| 2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one | |
| 2022 | |
| 225.2 | |
| Powder |
| 59277-89-3 | |
| White | |
| MFCD00057880 | |
| 14,146 | |
| Soluble in 1M HCl at 50mg/ml; Soluble in water at 0;7mg/ml; Also soluble in DMSO | |
| NC1=NC2=C(N=CN2COCCO)C(=O)N1 | |
| 225.21 | |
| CHEBI:2453 | |
| 98% |
Chemical Identifiers
| 59277-89-3 | |
| 225.21 | |
| MKUXAQIIEYXACX-UHFFFAOYSA-N | |
| 2022 | |
| 2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one |
| C8H11N5O3 | |
| MFCD00057880 | |
| acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum | |
| CHEBI:2453 | |
| NC1=NC2=C(N=CN2COCCO)C(=O)N1 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 261-685-1
RTECSNumber : UP0791400
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only