missing translation for 'onlineSavingsMsg'
Learn More

Acycloguanosine, 98%

Catalog No. AAJ6414403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6414403 1 g
AAJ6414406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6414403 Supplier Thermo Scientific Chemicals Supplier No. J6414403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits cytomegalovirus replication; induces apoptosis | CAS: 59277-89-3 | C8H11N5O3 | 225.21 g/mol

Inhibits cytomegalovirus replication; induces apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59277-89-3
Molecular Formula C8H11N5O3
Molecular Weight (g/mol) 225.21
MDL Number MFCD00057880
InChI Key MKUXAQIIEYXACX-UHFFFAOYSA-N
Synonym acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum
PubChem CID 2022
ChEBI CHEBI:2453
IUPAC Name 2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2COCCO)C(=O)N1

Specifications

Melting Point 256°C to 260°C
Color White
Quantity 1 g
Merck Index 14,146
Solubility Information Soluble in 1M HCl at 50mg/ml; Soluble in water at 0;7mg/ml; Also soluble in DMSO
Formula Weight 225.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Acycloguanosine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 261-685-1
RTECSNumber UP0791400
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only