missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACTR1B Recombinant Rabbit Monoclonal Antibody (23GB3135)

Catalog No. MA552597
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552597 20 μL
MA552596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552597 Supplier Invitrogen™ Supplier No. MA552597
Only null left

Rabbit Recombinant Monoclonal Antibody

ACTR1B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex.

Specifications

Antigen ACTR1B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3135
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Actr1b
Gene Accession No. P42025, Q8R5C5
Gene Alias 2310066K23Rik; AA960180; Actin-related protein 1B; Actr1b; AI851923; ARP1; ARP1 actin-related protein 1 homolog B, centractin beta; ARP1 actin-related protein 1 homolog B, centractin beta (yeast); ARP1 actin-related protein 1B, centractin beta; Arp1b; Beta-centractin; centractin beta; CTRN2; PC3; yeast homolog B
Gene Symbols Actr1b
Host Species Rabbit
Immunogen A synthetic peptide of human ACTR1B (301-376AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10120, 226977, 316333
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less