missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACTR1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114822
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114822 Supplier Invitrogen™ Supplier No. PA5114822
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACTR1A.

This gene encodes a 42.6 kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 8-13 copies per dynactin molecule, and is the most abundant molecule in the dynactin complex. It is an actin-related protein, and is approximately 60% identical at the amino acid level to conventional actin.

Specifications

Antigen ACTR1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACTR1A
Gene Accession No. P61163, P61164, P85515
Gene Alias actin related protein 1A; actin-related protein 1 alpha; actin-related protein 1 homolog A; actin-RPV; ACTR1A; actr1a {ECO:0000250; alpha-Arp1; alpha-centractin; ARP1; ARP1 actin related protein 1 homolog A; ARP1 actin-related protein 1 homolog A, centractin alpha; ARP1 actin-related protein 1 homolog A, centractin alpha (yeast); ARP1 actin-related protein 1A, centractin alpha; Arp1 centractin alpha; Arp1 contractin alpha; Arp1A; centractin; Centrosome-associated actin homolog; CTRN1; FLJ52695; FLJ52800; FLJ55002; UniProtKB:P61163}; yeast homolog A
Gene Symbols ACTR1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACTR1A(Accession P61163), corresponding to amino acid residues I180-K230.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10121, 294010, 54130
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less