missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin A Monoclonal Antibody (KAA378_4H12), Invitrogen™
Mouse Monoclonal Antibody
$1557.73 - $2745.56
Specifications
| Antigen | Activin A |
|---|---|
| Clone | KAA378_4H12 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
Description
Activin A Monoclonal antibody specifically detects Activin A in Human samples. It is validated for ELISA, Western BlotSpecifications
| Activin A | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product | |
| Recombinant human INHBA (proprietary range information). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Inhba |
| KAA378_4H12 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08476 | |
| 3624 | |
| IgG2b | |
| Protein A | |
| Antibody |