missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Powder, MilliporeSigma™ Supelco™

Catalog No. 111015406
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015406 250 g
111015405 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015406 Supplier Merck Emd Millipore Supplier No. 05105250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activated charcoal or activated carbon is an adsorbent used to remove broad spectrum of pollutants in air, water and soil, and is non-toxic in nature. It is odourless and tasteless and can be prepared from any carbonaceous material using gas or chemical activation methods. Gas activation is preferred when the raw material has less than 25% moisture content.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym Charcoal activated
IUPAC Name carbon
SMILES [C]

Specifications

Vapor Pressure <0.1 mmHg (20°C)
Physical Form Powder
Linear Formula C
Melting Point 3550°C
Quantity 250 g
Formula Weight 12.01