missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, For the Determination of AOX, 50-150 μm Particle Size, MilliporeSigma™ Supelco™

Catalog No. 111015407
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015407 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015407 Supplier Merck Emd Millipore Supplier No. 8048525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activated charcoal is a black powder that is generally obtained by subjecting wood or other carbonaceous materials to pyrolysis process.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym Charcoal activated
IUPAC Name carbon
SMILES [C]

Specifications

Vapor Pressure <0.1 mmHg (20°C)
Physical Form Powder
Linear Formula C
Melting Point 3550°C
Quantity 25 g
Formula Weight 12.01