missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Activated Charcoal, 150 g

Catalog No. 05405671
Change view
Click to view available options
Quantity:
150 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05405671 150 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05405671 Supplier BUCHI Supplier No. 11064971

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the adsorption stage of the scrubber, holds back particles and enables the aerosols to recondensate.

  • For the adsorption stage of the scrubber
  • Holds back particles
  • Enables aerosols to recondensate

Specifications

Chemical Name or Material Carbon
Quantity 150 g
Melting Point 3652°C
Packaging Glass or Plastic Bottle
Color Black
CAS 7440-44-0
Molecular Formula C
Synonym Graphite; Activated Charcoal
Molecular Weight (g/mol) 12
Percent Purity 100
Grade Technical
Show More Show Less