missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actinomycin D Crystalline MP Biomedicals
$174.60 - $695.16
Chemical Identifiers
| CAS | 50-76-0 |
|---|---|
| Molecular Formula | C62H86N12O16 |
| Molecular Weight (g/mol) | 1255.438 |
| MDL Number | MFCD00005033 |
| InChI Key | RJURFGZVJUQBHK-HUZQGMAJSA-N |
| Synonym | Dactinomycin, actinomycin-[thr-val-pro-sar-meval] |
| PubChem CID | 131954673 |
| IUPAC Name | 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, |
| SMILES | CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10465805
|
MP Biomedicals Inc
0210465805 |
5 mg |
Each for $174.60
|
|
|||||
|
ICN10465825
|
MP Biomedicals Inc
0210465825 |
25 mg |
Each for $695.16
|
|
|||||
Chemical Identifiers
| 50-76-0 | |
| 1255.438 | |
| RJURFGZVJUQBHK-HUZQGMAJSA-N | |
| 131954673 | |
| CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C |
| C62H86N12O16 | |
| MFCD00005033 | |
| Dactinomycin, actinomycin-[thr-val-pro-sar-meval] | |
| 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, |
Specifications
| Actinomycin D | |
| 50-76-0 | |
| 5 mg | |
| C62H86N12O16 | |
| Dactinomycin, actinomycin-[thr-val-pro-sar-meval] | |
| CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C | |
| 1255.438 | |
| 1255.5 |
| Crystalline Powder | |
| Red | |
| ∽ 98% | |
| MFCD00005033 | |
| RJURFGZVJUQBHK-HUZQGMAJSA-N | |
| 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, | |
| 131954673 | |
| Actinomycin D |
Safety and Handling
storageNote1 : Protect from light.
Recommended Storage : Store at 4°C.