missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actinomycin D, >99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210465880
Specifications
Actinomycin D | |
Powder | |
100 mg | |
C62H86N12O16 | |
RJURFGZVJUQBHK-HUZQGMAJSA-N | |
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, | |
131954673 |
Actinomycin D | |
50-76-0 | |
>99% | |
Dactinomycin, Actinomycin-[thr-val-pro-sar-meval] | |
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C | |
1255.438 | |
1255.5 |
Chemical Identifiers
50-76-0 | |
1255.438 | |
Dactinomycin, Actinomycin-[thr-val-pro-sar-meval] | |
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, |
C62H86N12O16 | |
RJURFGZVJUQBHK-HUZQGMAJSA-N | |
131954673 | |
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C |