missing translation for 'onlineSavingsMsg'
Learn More

Actinomycin D, >99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210465850

 View more versions of this product

Catalog No. ICN10465850


Only null left

Description

Description

Specifications

Specifications

Actinomycin D
Powder
50 mg
C62H86N12O16
RJURFGZVJUQBHK-HUZQGMAJSA-N
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
131954673
Actinomycin D
50-76-0
>99%
Dactinomycin, Actinomycin-[thr-val-pro-sar-meval]
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
1255.438
1255.5

Chemical Identifiers

50-76-0
1255.438
Dactinomycin, Actinomycin-[thr-val-pro-sar-meval]
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
C62H86N12O16
RJURFGZVJUQBHK-HUZQGMAJSA-N
131954673
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions