missing translation for 'onlineSavingsMsg'
Learn More

Acrylonitrile (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. A014625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A014625ML 25 mL
A0146500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A014625ML Supplier TCI America Supplier No. A014625ML

Chemical Identifiers

CAS 107-13-1
Molecular Formula C3H3N
Molecular Weight (g/mol) 53.064
MDL Number MFCD00001927
InChI Key NLHHRLWOUZZQLW-UHFFFAOYSA-N
Synonym acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox
PubChem CID 7855
ChEBI CHEBI:28217
IUPAC Name prop-2-enenitrile
SMILES C=CC#N

Specifications

Melting Point -84°C
Color Colorless
Boiling Point 79°C
Quantity 25 mL
UN Number 1093
Formula Weight 53.06
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Acrylonitrile (stabilized with MEHQ)
EINECSNumber (2)-1513
RTECSNumber AT5250000
TSCA Yes