accessibility menu, dialog, popup

UserName

CH2N

CAS RN 107-13-1

IUPAC Name: prop-2-enenitrile
Synonyms: acrylonitrile caswell no. 010 miller fumigrain acritet ccris 8 epa pesticide chemical code 000601 VCN nitrile acrilico cyanure de vinyle akrylonitryl
Molecular Weight (g/mol): 53.06
Molecular Formula: C3H3N
InChi Key: NLHHRLWOUZZQLW-UHFFFAOYSA-N
SMILES: C=CC#N
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)