missing translation for 'onlineSavingsMsg'
Learn More

Acrylonitrile, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019669
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019669 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019669 Supplier Merck Emd Millipore Supplier No. 315645ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylonitrile (AN) is a volatile liquid mostly used in manufacturing plastics and polymers.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 897°F
CAS 107-13-1
Vapor Pressure 86 mmHg (20°C)
Vapor Density 1.83 (vs. air)
Flash Point -5°C - Closed Cup
Physical Form Neat
Linear Formula CH2=CHCN
MDL Number MFCD00001927
Melting Point -83°C
Boiling Point 77°C
Synonym Vinyl cyanide
Type Analytical Standard
Quantity 5 mL
Formula Weight 53.06
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less