missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Acrylic Semi-Circular Block

With all faces fully polished.

Specifications

Product Type Semi-circular Block
Lens Material Acrylic
For Use With (Application) Light refraction and geometrical optics experiments
Diameter (English) Lens 3.5 in.
Diameter (Metric) Lens 90 mm
View More Specs

Products 2
Catalog Number Mfr. No. Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Width (Metric) Price Quantity & Availability  
S24386
View Documents Promotion Details
Eisco™
PH0570A
16 mm N/A
Only null left
 
S24387
View Documents Promotion Details
Eisco™
PH0570B
25 mm N/A
Only null left
 
Description

Description

  • Made from clear acrylic
Specifications

Specifications

Semi-circular Block
Light refraction and geometrical optics experiments
90 mm
Acrylic
3.5 in.
Videos
SDS
Documents

Documents