missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Acrylic Mirror Strips

Mirror strips ideal for optics demonstrations.

Supplier:  United Scientific™ MSA4X2

 View more versions of this product

Catalog No. S01319


Only null left
Explore available promotions

Description

Description

  • Made from Acrylic.
  • Mirror Strips are 3 mm thick.
Specifications

Specifications

4 x 2 in.
Acrylic
Mirror
3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions