missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Acrylic Mirror Strips

Mirror strips ideal for optics demonstrations.

Supplier:  United Scientific™ MSA6X4

 View more versions of this product

Catalog No. S01318

Only null left

Description

Description

  • Made from Acrylic.
  • Mirror Strips are 3 mm thick.
Specifications

Chemical Identifiers

Acrylic
Mirror
3 mm

Specifications

6 x 4 in.
Acrylic
Mirror
3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions