missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Acrylic Equilateral Prisms

Acrylic equilateral prisms are polished on all faces and designed for use in optics demonstrations or experiments.

$12.14 - $19.21

Specifications

Material Acrylic
For Use With (Application) Optics Demonstrations and Experiments
Product Type Prism
Shape Pyramid
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Focal Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Focal Price Quantity & Availability  
S27412
View Documents Promotion Details
Eisco™
PH0554AC
25 mm
Each for $12.14
Only null left
 
S27413
View Documents Promotion Details
Eisco™
PH0554CC
75 mm
Each for $19.21
Only null left
 
Description

Description

  • Made from high quality acrylic
  • Creates a rainbow when light passes through
  • Designed for use in educational optics experiments and demonstrations
  • Polished on all sides
  • Face Size: 25 mm
Specifications

Specifications

Acrylic
Optics Demonstrations and Experiments
Prism
Pyramid
Videos
SDS
Documents

Documents