missing translation for 'onlineSavingsMsg'
Learn More

Acrylic acid, 98%, extra pure, stabilized

Catalog No. AC164250025
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164250025 2.5 L Glass bottle
AC164252500 250 mL Glass bottle
AC164250010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164250025 Supplier Thermo Scientific Chemicals Supplier No. 164250025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79-10-7 | C3H4O2 | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-10-7 , 150-76-5
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00004367
InChI Key NIXOWILDQLNWCW-UHFFFAOYSA-N
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name prop-2-enoic acid
SMILES C=CC(=O)O

Specifications

CAS Min % 98
Melting Point 13.0°C
Density 1.0500g/mL
Boiling Point 139.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCO2H
Refractive Index 1.4192 to 1.4212
Quantity 2.5 L
Beilstein 02, 397
Specific Gravity 1.05
Merck Index 15, 121
Solubility Information Solubility in water: miscible. Other solubilities: miscible with most organic solvents,soluble in acetone
Stabilizer 180 to 220ppm MEHQ
Viscosity 1.3 mPa.s (20°C)
Formula Weight 72.06
Percent Purity 98%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Acrylic acid, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquat
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
EINECSNumber 201-177-9
Recommended Storage This product unavoidably polymerizes during storage

RUO – Research Use Only