missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 29:1, for biochemistry, 40% mix solution in water

Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol

$152.54 - $384.31

Specifications

Absorbance (1% in water), (1% in water) at 0nm, 0.1 max. at 290nm
Chemical Name or Material Acrylamide / N,N′-Methylenebisacrylamide 29:1
Physical Form Liquid
Acrylic Acid 0.001% max.
CAS 79-06-1 , 7732-18-5 , 110-26-9 , 79-06-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC330211000
View Documents
Thermo Scientific Chemicals
330211000
100 mL
Each for $152.54
Only null left
 
AC330215000
View Documents
Thermo Scientific Chemicals
330215000
500 mL
Each for $384.31
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
  • This compound polymerizes in the presence of free radicals to generate gels whose pore-size may be regulated by manipulating the relative concentrations of this mixture of compounds

Applications

  • Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
  • Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples
Specifications

Specifications

(1% in water), (1% in water) at 0nm, 0.1 max. at 290nm
Liquid
79-06-1 , 7732-18-5 , 110-26-9 , 79-06-1
61.0
Colorless
C3H5NO
HRPVXLWXLXDGHG-UHFFFAOYSA-N
prop-2-enamide
6579
Biochemical
Acrylamide / N,N′-Methylenebisacrylamide 29:1
0.001% max.
59.0
3μS/cm Max.
Glass Bottle
MFCD00008032
NC(=O)C=C
71.08
CHEBI:28619
Videos
SDS
Documents

Documents

RUO – Research Use Only