missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide-d3 Standard Solution, ∼500 mg/L in Acetonitrile, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 122775-19-3 |
| Flash Point | 2°C - Closed Cup |
| Type | Analytical Standard |
| Quantity | 5 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | ̃500 mg/L in acetonitrile |