missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Acrylamide/Bisacrylamide 37.5:1, powder
Premixed powder for preparing nucleic acid PAGE gels
Specifications
| Chemical Name or Material | Acrylamide/Bisacrylamide 37.5:1 |
|---|---|
| MDL Number | MFCD00080848 |
| UN Number | UN2074 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6122017
|
Thermo Scientific™
J6122017 |
40g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AAJ6122028
|
Thermo Scientific™
J6122028 |
200g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Acrylamide/Bisacrylamide 37.5:1 | |
| MFCD00080848 | |
| UN2074 |
RUO â Research Use Only