missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Acrylamide/Bisacrylamide 37.5:1, powder
Premixed powder for preparing nucleic acid PAGE gels
Supplier: Thermo Scientific™ J6122017
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Acrylamide/Bisacrylamide 37.5:1 | |
| 40g | |
| UN2074 |
| White | |
| MFCD00080848 | |
| Soluble in water |
RUO â Research Use Only