Learn More
Thermo Scientific Chemicals Acrylamide (30%) Solutions
CAS: 79-06-1 | C3H5NO | 71.08 g/mol
$226.33 - $419.58
Chemical Identifiers
| CAS | 79-06-1 |
|---|---|
| Molecular Formula | C3H5NO |
| Molecular Weight (g/mol) | 71.08 |
| MDL Number | MFCD00008032 |
| InChI Key | HRPVXLWXLXDGHG-UHFFFAOYSA-N |
| PubChem CID | 6579 |
| ChEBI | CHEBI:28619 |
| IUPAC Name | prop-2-enamide |
| SMILES | NC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ62100AP
|
Thermo Scientific Chemicals
J62100AP |
500 mL |
Each for $226.33
|
|
|||||
|
AAJ62100K2
|
Thermo Scientific Chemicals
J62100K2 |
1 L |
Each for $419.58
|
|
|||||
Description
Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from Acids, Oxidizing agents, Iron and iron salts., Copper, Brass, Free radical initiators.
Chemical Identifiers
| 79-06-1 | |
| 71.08 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| CHEBI:28619 | |
| NC(=O)C=C |
| C3H5NO | |
| MFCD00008032 | |
| 6579 | |
| prop-2-enamide |
Specifications
| Liquid | |
| 30% soln | |
| 500 mL | |
| MFCD00008032 | |
| 605349 | |
| Soluble in water. | |
| NC(=O)C=C | |
| 71.08 | |
| CHEBI:28619 |
| 79-06-1 | |
| Colorless | |
| C3H5NO | |
| UN3426 | |
| 14,129 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| prop-2-enamide | |
| 6579 | |
| Acrylamide, bisacrylamide free |
Safety and Handling
GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H301-H315-H317-H319-H332-H340-H350-H361f-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
EINECSNumber : 201-173-7
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only