missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide (30%) Solutions

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

Supplier:  Thermo Scientific Chemicals J62100K2

 View more versions of this product

Catalog No. AAJ62100K2


Only null left

Description

Description

Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from Acids, Oxidizing agents, Iron and iron salts., Copper, Brass, Free radical initiators.
Specifications

Specifications

Acrylamide
Liquid
30% soln
1 L
MFCD00008032
605349
Soluble in water.
NC(=O)C=C
71.08
CHEBI:28619
bisacrylamide free
79-06-1
Colorless
C3H5NO
UN3426
14,129
HRPVXLWXLXDGHG-UHFFFAOYSA-N
prop-2-enamide
6579

Chemical Identifiers

79-06-1
71.08
HRPVXLWXLXDGHG-UHFFFAOYSA-N
CHEBI:28619
NC(=O)C=C
C3H5NO
MFCD00008032
6579
prop-2-enamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c

H301-H315-H317-H319-H332-H340-H350-H361f-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION

EINECSNumber : 201-173-7

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only