missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acrosomal Vesicle Protein Recombinant Rabbit Monoclonal Antibody (23GB4285)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552675
Description
Acrosomal Vesicle Protein Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. This gene consists of 4 exons and its alternative splicing generates multiple distinct transcripts, which encode protein isoforms ranging from 81 to 265 amino acids. The longest transcript is the most abundant, comprising 53-72% of the total acrosomal vesicle protein 1 messages; the second largest transcript comprises 15-32%; the third and the fourth largest transcripts account for 3.4-8.3% and 8.7-12.5%, respectively; and the remaining transcripts combined account for < 1% of the total acrosomal vesicle protein 1 message. It is suggested that phenomena of cryptic splicing and exon skipping occur within this gene. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration, and it is a potential contraceptive vaccine immunogen for humans.Chemical Identifiers
| Acrosomal Vesicle Protein | |
| 1.2 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26436 | |
| 56 | |
| A synthesized peptide derived from human Intra Acrosomal Protein (200-265AA). | |
| Antibody |
| 23GB4285 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| ACRV1 | |
| acrosomal protein SP-10; Acrosomal vesicle protein 1; Acrv1; D11S4365; Msa63; MSA-63; rSP10; Sp10; SP-10; SPACA2; sperm membrane protein; sperm protein 10 | |
| ACRV1 | |
| Primary | |
| Affinity chromatography |
Specifications
| Acrosomal Vesicle Protein | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26436 | |
| ACRV1 | |
| A synthesized peptide derived from human Intra Acrosomal Protein (200-265AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 23GB4285 | |
| Unconjugated | |
| ACRV1 | |
| acrosomal protein SP-10; Acrosomal vesicle protein 1; Acrv1; D11S4365; Msa63; MSA-63; rSP10; Sp10; SP-10; SPACA2; sperm membrane protein; sperm protein 10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56 | |
| -20°C | |
| Liquid |