missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein and Acrylonitrile in Methanol Purge and Trap, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC 603XM
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Acrolein | 107-02-8 | 2000μg/mL |
| Acrylonitrile | 107-13-1 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Acrolein and Acrylonitrile in Methanol P&T | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| AA, ICP | |
| Certified Reference Material | |
| 1 mL |
| 2mL amber prescored glass ampule | |
| For US EPA Method 603 | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| Methanol-P and T | |
| Components at 2000μg/mL in Methanol: Acrolein; Acrylonitrile |
Chemical Identifiers
| 0.0 |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only