missing translation for 'onlineSavingsMsg'
Learn More

Acriflavine (mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine) 70.0+%, TCI America™

Supplier:  TCI America A013425G

Catalog No. A013425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acriflavine (mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine)
C27H25ClN6
25 g
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
468.99
Crystalline Powder
8048-52-0
MFCD00064307
PEJLNXHANOHNSU-UHFFFAOYSA-N
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
≥70.0% (HPLC)

Chemical Identifiers

8048-52-0
468.99
PEJLNXHANOHNSU-UHFFFAOYSA-N
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
C27H25ClN6
MFCD00064307
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AR9660000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions