missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACRC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149609
Description
Immunogen sequence: EDKSDDSDVP DDNSDDLEVP VPAEDLCNEG QIASDEEELV EAAAAVSQHD SSDDAGEQDL GENLSKPPSD PEANPEVSER KLPTE Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 30%.
Probable catalytic subunit of the acryloyl-CoA reductase complex involved in the pathway of L-alanine fermentation. Catalyzes the irreversible NADH-dependent formation of propionyl-CoA from acryloyl-CoA. It can also use 3-buten-2-one as substrate.Chemical Identifiers
| ACRC | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| GCNA | |
| acidic repeat containing; acidic repeat-containing protein; ACRC; GCNA; Germ cell nuclear antigen; NAAR1; putative nuclear protein | |
| GCNA | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96QF7 | |
| 93953 | |
| Recombinant protein corresponding to Human GCNA. | |
| Antibody |
Specifications
| ACRC | |
| Polyclonal | |
| Unconjugated | |
| GCNA | |
| acidic repeat containing; acidic repeat-containing protein; ACRC; GCNA; Germ cell nuclear antigen; NAAR1; putative nuclear protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93953 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96QF7 | |
| GCNA | |
| Recombinant protein corresponding to Human GCNA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |